ContactUsLinkComponent
GC capillary column ROTI®CAP-WAX Polyethyleneglycol (PEG) 20000 Daltons • Polar • recommended for the solvent analytics of alcohols • suitable for aequous solutions • high temperature stability Length:,,30 m Ø interior 0,25 mm layer thickness 0,25 µm 1 * 1 items
  ROTHCN12.1
 :  Roth Carl
GC capillary column ROTI®CAP-WAX Polyethyleneglycol (PEG) 20000 Daltons • Polar • recommended for the solvent analytics of alcohols • suitable for aequous solutions • high temperature stability Length:,,30 m Ø interior 0,25 mm layer thickness 0,25 µm 1 * 1 items
  ROTHCN12.1
 :  Roth Carl
Frequently Bought Together