Double bosshead
Double bosshead with M8 brass thumb bolt, 16 mm
Catalog # JUCHVL04810
Supplier: JUCHHEIM Laborgerate GmbH
Jump to:
Product Details & Documents
Double bosshead made from epoxy coated malleable iron.
Certifications: DIN 12895
Specifications
- Description:Double bosshead with M8 brass thumb bolt
- Seq:01
- For rod Ø:16 mm