Aluminium oxide activated basic HPLC flash grade
250 g
Supplier: Thermo Fisher Scientific
Aluminium oxide activated basic HPLC flash grade
Catalog # 45949.30
Supplier: Thermo Fisher Scientific
CAS Number: 1344-28-1
Jump to:
Product Details & Documents
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 18-32 Micron APS Powder Refractive Index: 1.765 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424
- CAS Number:1344-28-1
- Formula:Al₂O₃
- MDL Number:MFCD00003424
- Molecular Weight:101.96 g/mol
- Merck Index:14,356
- Boiling Pt:2980 °C
- Melting Pt:2045 °C
- Storage Temperature:Ambient
- EINECS:215-691-6
Documents
Specifications
Specifications
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



