Aluminium oxide activated basic HPLC flash grade
1 kg
Supplier: Thermo Fisher Scientific
Aluminium oxide activated basic HPLC flash grade
Catalog # 45949.A1
Supplier: Thermo Fisher Scientific
CAS Number: 1344-28-1
Jump to:
Product Details & Documents
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 18-32 Micron APS Powder Refractive Index: 1.765 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424
- CAS Number:1344-28-1
- Formula:Al₂O₃
- MDL Number:MFCD00003424
- Molecular Weight:101.96 g/mol
- Merck Index:14,356
- Boiling Pt:2980 °C
- Melting Pt:2045 °C
- Storage Temperature:Ambient
- EINECS:215-691-6
Documents
Specifications
Specifications
- Pk:1 kg



