Orders LinkContactUsLinkComponent
Blocking buffer, fish gelatin
Blocking buffer, fish gelatin
Fish gelatin powder, 2×50 µg, 100 g
Catalog # 22011.
Supplier:   Biotium
Blocking buffer, fish gelatin
Catalog # 22011.
Supplier:  Biotium
CAS Number:  
Blocking buffer, fish gelatin
Fish gelatin powder, 2×50 µg, 100 g
CAS Number:  
Price
SFr. 82.90
Log in for availability
1 * 100 g
Product Details & Documents
Fish gelatin is commonly used in blocking solutions to minimise background noise in immunodetection procedures such as Western Blotting and immunostaining. Fish gelatin can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or for blocking and dilution of antibodies during immunostaining procedures. Unlike BSA or milk, fish gelatin does not contain IgG or serum proteins that could cross-react with mammalian antibodies.
Specifications
Specifications
  • Pk:
    100 g
  • Description:
    Fish gelatin powder, 2×50 µg
Specifications