Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Tanks, rectangular, LDPE, LaboPlast®

250 L, Seamless containers, rectangular shaped, 1000×600×450 mm
Codice catalogo BURK3440-2506
Nome del fornitore:   BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Codice catalogo BURK3440-2506
Nome del fornitore:  BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®

250 L, Seamless containers, rectangular shaped, 1000×600×450 mm

Price
SFr. 805.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.

  • Linear LDPE, transparent
  • Rupture-proof
Specifiche
  • Capacity:
    250 L
  • Description:
    Seamless containers, rectangular shaped
  • Seq:
    15
  • W×D×H:
    1000×600×450 mm
Accessories

Informazioni sul prodotto

  • Codice catalogo
    Description
    Disponibilità
    Price
  • BURK3440-0034
    Container thread ¾" for cylindrical containers
    Log in per disponibilità
    1 * 1 ST - SFr. 202.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.