Cryoboxes
9×9, 12,8 mm, 133×133×30 mm
Catalog # RATI5220103
Supplier: RATIOLAB
Jump to:
Product Details & Documents
These cryoboxes are made of polypropylene in natural or other colors. They can be used with sample vessels of differing diameters by removing the grid inserts.
- Robust hinge with safe snap-on lid
- Stackable for safe transport
- Drain bores at the bottom for dew liquids
These cryoboxes are particularly practical and flexible due to their removable inserts. Cleaning of the boxes and the inserts is much simpler than cryoboxes with permanently fitted inserts.
The inserts fit all ratiolab® cryo boxes.
Lieferinformationen: Boxes are supplied without inserts, they need to be ordered separately.
Specifications
- Compartments:9×9
- For tube Ø:12,8 mm
- W×D×H ext.:133×133×30 mm
- Seq:15
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



