Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dropping funnels, cylindrical, with double jacket

Dropping funnels, cylindrical, with double jacket

Dropping funnel with drilling 2,5 mm, 100 ml, 19/26 NS
Catalog # NEUB1596-20-0119
Dropping funnels, cylindrical, with double jacket
Catalog # NEUB1596-20-0119
Dropping funnels, cylindrical, with double jacket

Dropping funnel with drilling 2,5 mm, 100 ml, 19/26 NS

Price
SFr. 697.00
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3, with PTFE tap.

  • Graduated
  • With drop counter
  • with GL 14 olives (PE)
  • With pressure compensation
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml
  • Description:
    Dropping funnel with drilling 2,5 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.