Pyrocatechol ≥99%
250 g
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Catechol, 99% 1 * 250 g Catalog #A10164.30
Product A10164.30 is a replacement for ABCRAB108846.0250 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Purity: 99% CAS No.: 120-80-9 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 127°(260°F) Physical Form: Powder or flakes Density: 1.370 Sensitivity: Air and Light Sensitive Appearance: White to brown Odour: Aromatic Melting Point: 104-107° Boiling Point: 245-246° Beilstein Reference: 471401 Merck Reference: 14,7999 UN No.: 2811 MDL No.: MFCD00002188
Synonyms: o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
- CAS Number:120-80-9
- Formula:C₆H₆O₂
- MDL Number:MFCD00002188
- Molecular Weight:110.11 g/mol
- Merck Index:14,7999
- Boiling Pt:245…246 °C
- Melting Pt:104…107 °C
- Density:1.370
- Flash Pt:127 °C (260 °F)
- Storage Temperature:Ambient
- EINECS:204-427-5
- UN:2811
- ADR:6.1,III
Documents
Specifications
Specifications
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



