TFFH Tetramethylfluoroformamidinium hexafluorophosphate 1 * 5 g
Catalog # 8.51090.0005
Supplier: Novabiochem (Part of Merck)
TFFH Tetramethylfluoroformamidinium hexafluorophosphate 1 * 5 g
Catalog # 8.51090.0005
Supplier: Novabiochem (Part of Merck)
Frequently Bought Together
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Supplier: Merck
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Mineral oil light, white NF, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals