Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Accessories

Accessories

Glass tube, 200 mm, 15 ml
Catalog # KRUEPRG-200
Supplier:   A.KRÜSS Optronic
Accessories
Catalog # KRUEPRG-200
Supplier:  A.KRÜSS Optronic
Accessories

Glass tube, 200 mm, 15 ml

Price
SFr. 333.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Glass tube, 200 mm, 15 ml
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.