Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Jerricans, polyethlene

Jerricans, polyethlene

10 l, 298×198×264 mm, Black
Catalog # DUEH5.11.010
Supplier:   DUPERTHAL
Jerricans, polyethlene
Catalog # DUEH5.11.010
Supplier:  DUPERTHAL
Jerricans, polyethlene
Jerricans, polyethlene

10 l, 298×198×264 mm, Black

Price
SFr. 347.00
Log in for availability
1 * 1 ST
Product Details & Documents

PE, electrostatically conductive.

  • Good chemical resistance
  • Good temperature resistance
  • Recyclable
Specifications
  • Capacity:
    10 l
  • Colour:
    Black
  • W×D×H:
    298×198×264 mm
Accessories

Product Information

  • Catalog #
    For
    Ø
    Availability
    Price
  • Safety funnel
    Safety funnel
    DUEH5.08.001
    Flammable liquids
    120 mm
    Log in for availability
    1 * 1 ST - SFr. 220.00

Product Information

  • Catalog #
    Availability
    Price
  • Special pliers
    Special pliers
    DUEH5.09.003
    Log in for availability
    1 * 1 ST - SFr. 9.60