Peqlab peqGOLD, Deoxynucleotide Triphosphate (Solution)
dATP, 100 mM, 25 µmol, 250 µl, 1
Supplier: VWR Collection
Peqlab peqGOLD, Deoxynucleotide Triphosphate (Solution)
Catalog # 732-3155
Supplier: VWR Collection
CAS Number:
Jump to:
Product Details & Documents
Individually packaged deoxynucleotide triphosphates manufactured to the highest quality levels.
- In solution and ready to use, pH 7,5
- Synthetically manufactured
- Purity, measured by HPLC, >99%
- Highly stable, >90% stability after 6 weeks storage at room temperature
: Manufacturing and quality control under ISO 9001 and ISO 14001.
Specifications
Specifications
- Pk:1
- Description:dATP, 100 mM, 25 µmol, 250 µl



