ContactUsLinkComponent
40 assays per kit. For the specific assay of L-ascorbic acid in beverages, meat, flour, dairy and vegetable products. 1 * 1 KIT
Catalog # 710-1954
Supplier:  MEGAZYME
40 assays per kit. For the specific assay of L-ascorbic acid in beverages, meat, flour, dairy and vegetable products. 1 * 1 KIT
Catalog # 710-1954
Supplier:  MEGAZYME
Frequently Bought Together