Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Student standard pattern forceps

Student standard pattern forceps

Stainless steel, 160 mm, Serrated, Straight, 3,5 mm
Catalog # FSCI91100-16
Marketsource
Supplier:   FINE SCIENCE TOOLS
Student standard pattern forceps
Catalog # FSCI91100-16
Supplier:  FINE SCIENCE TOOLS
Marketsource
Student standard pattern forceps
Student standard pattern forceps

Stainless steel, 160 mm, Serrated, Straight, 3,5 mm

Price
SFr. 12.83
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Length:
    160 mm
  • Material:
    Stainless steel
  • Seq:
    03
  • Tip style:
    Serrated
  • Tip width:
    3,5 mm
  • Tip shape:
    Straight