Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Schwartz micro serrefines

Schwartz micro serrefines

Strong clamp press, Stainless steel, 26 mm, Slightly curved, 10×1,75 mm
Codice catalogo FSCI18052-02
Marketsource
Nome del fornitore:   FINE SCIENCE TOOLS
Schwartz micro serrefines
Codice catalogo FSCI18052-02
Nome del fornitore:  FINE SCIENCE TOOLS
Marketsource
Schwartz micro serrefines
Schwartz micro serrefines

Strong clamp press, Stainless steel, 26 mm, Slightly curved, 10×1,75 mm

Price
11.45 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Specifiche
  • Description:
    Strong clamp press
  • Length:
    26 mm
  • Material:
    Stainless steel
  • Seq:
    03
  • Tip size:
    10×1,75 mm
  • Tip shape:
    Slightly curved
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.