Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Schwartz micro serrefines

Strong clamp press, Stainless steel, 26 mm, Curved, 10×1,75 mm
  FSCI18052-03
Marketsource
Schwartz micro serrefines
  FSCI18052-03
Marketsource
Schwartz micro serrefines
Schwartz micro serrefines

Strong clamp press, Stainless steel, 26 mm, Curved, 10×1,75 mm

 
10.54 SFr.
   
1 * 1 ST
 
 
  • Description:
    Strong clamp press
  • Length:
    26 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip size:
    10×1,75 mm
  • Tip shape:
    Curved
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.