Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Pearson rongeurs

Pearson rongeurs, 1,2 mm cup diameter, Shallow bend, 190 mm
  FSCI16015-17
Marketsource
Pearson rongeurs
  FSCI16015-17
Marketsource
Pearson rongeurs
Pearson rongeurs

Pearson rongeurs, 1,2 mm cup diameter, Shallow bend, 190 mm

 
SFr. 251.40
   
1 * 1 ST
 

These rongeurs are made of stainless steel.

  • Single joint action
 
  • Type:
    Shallow bend
  • Description:
    Pearson rongeurs, 1,2 mm cup diameter
  • Length:
    190 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.