Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Tissue forceps, 1×2 teeth

Stainless steel, 145 mm, 1×2 teeth, Straight, 2×1,5 mm
  FSCI11021-14
Marketsource
Tissue forceps, 1×2 teeth
  FSCI11021-14
Marketsource
Tissue forceps, 1×2 teeth
Tissue forceps, 1×2 teeth

Stainless steel, 145 mm, 1×2 teeth, Straight, 2×1,5 mm

 
SFr. 14.66
   
1 * 1 ST
 
 
  • Length:
    145 mm
  • Material:
    Stainless steel
  • Seq:
    03
  • Tip style:
    1×2 teeth
  • Tip size:
    2×1,5 mm
  • Tip shape:
    Straight