Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Tissue forceps, 2×3 teeth

Stainless steel, 145 mm, 2x3 teeth, Straight, 2,9×2 mm
  FSCI11022-14
Marketsource
Tissue forceps, 2×3 teeth
  FSCI11022-14
Marketsource
Tissue forceps, 2×3 teeth
Tissue forceps, 2×3 teeth

Stainless steel, 145 mm, 2x3 teeth, Straight, 2,9×2 mm

 
SFr. 25.19
   
1 * 1 ST
 
 
  • Length:
    145 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip style:
    2x3 teeth
  • Tip size:
    2,9×2 mm
  • Tip shape:
    Straight