Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Tissue forceps, slim 1×2 teeth

Stainless steel, 155 mm, 1×2 teeth, Straight, 1,7×1,3 mm
  FSCI11023-15
Marketsource
Tissue forceps, slim 1×2 teeth
  FSCI11023-15
Marketsource
Tissue forceps, slim 1×2 teeth
Tissue forceps, slim 1×2 teeth

Stainless steel, 155 mm, 1×2 teeth, Straight, 1,7×1,3 mm

 
SFr. 19.24
   
1 * 1 ST
 
 
  • Length:
    155 mm
  • Material:
    Stainless steel
  • Seq:
    04
  • Tip style:
    1×2 teeth
  • Tip size:
    1,7×1,3 mm
  • Tip shape:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.