Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Wagner scissors

Wagner scissors

Sharp/sharp, curved tips, 120 mm, 30 mm
Catalog # FSCI14069-12
Marketsource
Supplier:   FINE SCIENCE TOOLS
Wagner scissors
Catalog # FSCI14069-12
Supplier:  FINE SCIENCE TOOLS
Marketsource
Wagner scissors
Wagner scissors

Sharp/sharp, curved tips, 120 mm, 30 mm

Price
SFr. 53.13
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Sharp/sharp, curved tips
  • Length:
    120 mm
  • Seq:
    01
  • Blade length:
    30 mm