Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Potts-Smith forceps

Stainless steel, 180 mm, Serrated, Straight, 1,25×1 mm
  FSCI11012-18
Marketsource
Potts-Smith forceps
  FSCI11012-18
Marketsource
Potts-Smith forceps
Potts-Smith forceps

Stainless steel, 180 mm, Serrated, Straight, 1,25×1 mm

 
SFr. 22.44
   
1 * 1 ST
 
 
  • Length:
    180 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip style:
    Serrated
  • Tip size:
    1,25×1 mm
  • Tip shape:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.