Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Fine scissors, extra large loops

Sharp/sharp, straight tips, large loops, 95 mm, 14 mm
  FSCI14106-09
Marketsource
Fine scissors, extra large loops
  FSCI14106-09
Marketsource
Fine scissors, extra large loops
Fine scissors, extra large loops

Sharp/sharp, straight tips, large loops, 95 mm, 14 mm

 
SFr. 56.80
   
1 * 1 ST
 

These scissors are made of stainless steel.

 
  • Type:
    Sharp/sharp, straight tips, large loops
  • Length:
    95 mm
  • Seq:
    01
  • Blade length:
    14 mm