Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Mayo scissors

Blunt/blunt, straight tips, Toughcut®, 150 mm, 39 mm, +
  FSCI14110-15
Marketsource
Mayo scissors
  FSCI14110-15
Marketsource
Mayo scissors
Mayo scissors

Blunt/blunt, straight tips, Toughcut®, 150 mm, 39 mm, +

 
SFr. 57.71
   
1 * 1 ST
 

These scissors are made of stainless steel.

 
  • Type:
    Blunt/blunt, straight tips, Toughcut®
  • Length:
    150 mm
  • Seq:
    01
  • Serrated:
    +
  • Blade length:
    39 mm