Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Mayo scissors

Blunt/blunt, curved tips, Toughcut®, 150 mm, 39 mm, +
Catalog # FSCI14111-15
Marketsource
Supplier:   FINE SCIENCE TOOLS
Mayo scissors
Catalog # FSCI14111-15
Supplier:  FINE SCIENCE TOOLS
Marketsource
Mayo scissors
Mayo scissors

Blunt/blunt, curved tips, Toughcut®, 150 mm, 39 mm, +

Price
SFr. 61.38
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Blunt/blunt, curved tips, Toughcut®
  • Length:
    150 mm
  • Seq:
    02
  • Serrated:
    +
  • Blade length:
    39 mm