Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Strabismus scissors

Strabismus scissors

Blunt/blunt, curved tips, 90 mm, 19 mm
Catalog # FSCI14075-09
Marketsource
Supplier:   FINE SCIENCE TOOLS
Strabismus scissors
Catalog # FSCI14075-09
Supplier:  FINE SCIENCE TOOLS
Marketsource
Strabismus scissors
Strabismus scissors

Blunt/blunt, curved tips, 90 mm, 19 mm

Price
SFr. 61.84
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Blunt/blunt, curved tips
  • Length:
    90 mm
  • Seq:
    01
  • Blade length:
    19 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.