Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Extra fine Graefe forceps

Extra fine Graefe forceps

Stainless steel, 100 mm, 1×2 teeth, Curved, 0,5×0,5 mm, 0,5 mm
  FSCI11155-10
Marketsource
Extra fine Graefe forceps
  FSCI11155-10
Marketsource
Extra fine Graefe forceps
Extra fine Graefe forceps

Stainless steel, 100 mm, 1×2 teeth, Curved, 0,5×0,5 mm, 0,5 mm

 
SFr. 58.17
   
1 * 1 ST
 
 
  • Length:
    100 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip style:
    1×2 teeth
  • Tip size:
    0,5×0,5 mm
  • Tip width:
    0,5 mm
  • Tip shape:
    Curved
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.