Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ring forceps

Ring forceps

Stainless steel, 90 mm, Smooth, Straight, 6×0,9 mm
Catalog # FSCI11106-09
Marketsource
Supplier:   FINE SCIENCE TOOLS
Ring forceps
Catalog # FSCI11106-09
Supplier:  FINE SCIENCE TOOLS
Marketsource
Ring forceps
Ring forceps

Stainless steel, 90 mm, Smooth, Straight, 6×0,9 mm

Price
SFr. 52.22
Log in for availability
1 * 1 ST
Product Details & Documents

For transport of tissue and organs. A limit screw prevents excessive applied pressure.

Specifications
  • Length:
    90 mm
  • Material:
    Stainless steel
  • Seq:
    03
  • Tip style:
    Smooth
  • Tip size:
    6×0,9 mm
  • Tip shape:
    Straight