Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Live insect forceps

Live insect forceps

Stainless steel, 100 mm, Sharp, Straight, 0,9×0,15 mm
Catalog # FSCI26029-10
Marketsource
Supplier:   FINE SCIENCE TOOLS
Live insect forceps
Catalog # FSCI26029-10
Supplier:  FINE SCIENCE TOOLS
Marketsource
Live insect forceps
Live insect forceps

Stainless steel, 100 mm, Sharp, Straight, 0,9×0,15 mm

Price
SFr. 22.44
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Length:
    100 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip style:
    Sharp
  • Tip size:
    0,9×0,15 mm
  • Tip shape:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.