Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Micro Curettes

Micro Curettes

Micro curette, tip diameter 1,0 mm, 130 mm
Catalog # FSCI10081-10
Marketsource
Supplier:   FINE SCIENCE TOOLS
Micro Curettes
Catalog # FSCI10081-10
Supplier:  FINE SCIENCE TOOLS
Marketsource
Micro Curettes

Micro curette, tip diameter 1,0 mm, 130 mm

Price
SFr. 76.04
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. A curette tip consists of a scoop with sharp edges and is used for scraping or cleaning. It also can be used for separating and transporting, inoculating plates or transferring small amounts of chemicals.

Specifications
  • Description:
    Micro curette, tip diameter 1,0 mm
  • Length:
    130 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.