Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Surgical scissors, sharp

Sharp/sharp, straight tips, 130 mm, 42 mm
  FSCI14002-13
Marketsource
Surgical scissors, sharp
  FSCI14002-13
Marketsource
Surgical scissors, sharp
Surgical scissors, sharp

Sharp/sharp, straight tips, 130 mm, 42 mm

 
SFr. 31.61
   
1 * 1 ST
 

These scissors are made of stainless steel.

 
  • Type:
    Sharp/sharp, straight tips
  • Length:
    130 mm
  • Seq:
    04
  • Blade length:
    42 mm