Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Surgical scissors, sharp

Surgical scissors, sharp

Sharp/sharp, curved tips, 120 mm, 41 mm
Catalog # FSCI14005-12
Marketsource
Supplier:   FINE SCIENCE TOOLS
Surgical scissors, sharp
Catalog # FSCI14005-12
Supplier:  FINE SCIENCE TOOLS
Marketsource
Surgical scissors, sharp
Surgical scissors, sharp

Sharp/sharp, curved tips, 120 mm, 41 mm

Price
SFr. 34.35
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Sharp/sharp, curved tips
  • Length:
    120 mm
  • Seq:
    01
  • Blade length:
    41 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.