Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Metzenbaum scissors | Avantor

Metzenbaum scissors

Sharp/blunt, straight tips, 145 mm, 31 mm
Katalognr. FSCI14024-14
Marketsource
Anbieter:   FINE SCIENCE TOOLS
Metzenbaum scissors
Katalognr. FSCI14024-14
Anbieter:  FINE SCIENCE TOOLS
Marketsource
Metzenbaum scissors
Metzenbaum scissors

Sharp/blunt, straight tips, 145 mm, 31 mm

Preis
SFr. 50.39
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Technische Daten
  • Type:
    Sharp/blunt, straight tips
  • Length:
    145 mm
  • Seq:
    03
  • Blade length:
    31 mm