Fine scissors, sharp
Sharp/sharp, curved tips to side, 105 mm, 21 mm
FSCI14062-10
Marketsource
These scissors are made of stainless steel.
- Type:Sharp/sharp, curved tips to side
- Length:105 mm
- Seq:03
- Blade length:21 mm
These scissors are made of stainless steel.