Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stevens Tenotomy scissors

Stevens Tenotomy scissors

Sharp/sharp, curved tips, 110 mm, 21 mm
Catalog # FSCI14065-11
Marketsource
Supplier:   FINE SCIENCE TOOLS
Stevens Tenotomy scissors
Catalog # FSCI14065-11
Supplier:  FINE SCIENCE TOOLS
Marketsource
Stevens Tenotomy scissors
Stevens Tenotomy scissors

Sharp/sharp, curved tips, 110 mm, 21 mm

Price
SFr. 61.84
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Sharp/sharp, curved tips
  • Length:
    110 mm
  • Seq:
    03
  • Blade length:
    21 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.