Fine scissors, sharp
Sharp/sharp, straight tips, 105 mm, 25 mm
FSCI14160-10
Marketsource
These scissors are made of stainless steel.
- Type:Sharp/sharp, straight tips
- Length:105 mm
- Seq:09
- Blade length:25 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



