Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Fine scissors, sharp

Sharp/sharp, straight tips, 105 mm, 25 mm
  FSCI14161-10
Marketsource
Fine scissors, sharp
  FSCI14161-10
Marketsource
Fine scissors, sharp
Fine scissors, sharp

Sharp/sharp, straight tips, 105 mm, 25 mm

 
SFr. 84.28
   
1 * 1 ST
 

These scissors are made of stainless steel.

 
  • Type:
    Sharp/sharp, straight tips
  • Length:
    105 mm
  • Seq:
    10
  • Blade length:
    25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.