Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Mayo-Stille scissors

Mayo-Stille scissors

Blunt/blunt, curved tips, 170 mm, 45 mm
Catalog # FSCI14013-17
Marketsource
Supplier:   FINE SCIENCE TOOLS
Mayo-Stille scissors
Catalog # FSCI14013-17
Supplier:  FINE SCIENCE TOOLS
Marketsource
Mayo-Stille scissors
Mayo-Stille scissors

Blunt/blunt, curved tips, 170 mm, 45 mm

Price
SFr. 48.10
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Documents

Specifications
  • Type:
    Blunt/blunt, curved tips
  • Length:
    170 mm
  • Seq:
    03
  • Blade length:
    45 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.