Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Mayo-Noble scissors

Mayo-Noble scissors

Blunt/blunt, curved tips, 165 mm, 37 mm
Catalog # FSCI14015-17
Marketsource
Supplier:   FINE SCIENCE TOOLS
Mayo-Noble scissors
Catalog # FSCI14015-17
Supplier:  FINE SCIENCE TOOLS
Marketsource
Mayo-Noble scissors
Mayo-Noble scissors

Blunt/blunt, curved tips, 165 mm, 37 mm

Price
SFr. 76.04
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Blunt/blunt, curved tips
  • Length:
    165 mm
  • Seq:
    01
  • Blade length:
    37 mm