Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Metzenbaum scissors

Metzenbaum scissors

Blunt/blunt, curved tips, 180 mm, 35 mm
  FSCI14017-18
Marketsource
Metzenbaum scissors
  FSCI14017-18
Marketsource
Metzenbaum scissors
Metzenbaum scissors

Blunt/blunt, curved tips, 180 mm, 35 mm

 
SFr. 50.84
   
1 * 1 ST
 

These scissors are made of stainless steel.

 
  • Type:
    Blunt/blunt, curved tips
  • Length:
    180 mm
  • Seq:
    04
  • Blade length:
    35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.