Shodex GPC KF-2002.5 - polymer based organic HPLC column - SEC mode - THF - preparative - ID: 20mm length: 300mm - Functional Group: - - Base Material: Styrene dinvinylbenzene copolymer - Particle Size (µm): 6 - Pore Size (A): - - Exclusion Limit (g 1 * 1 items
Catalog # SHOWF6102425
Supplier: Resonac
Frequently Bought Together