Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Plasmid Maxiprep kit, GeneJET™

Plasmid Maxiprep kit, GeneJET™

GeneJET™ Plasmid Maxiprep Kit, 25 preps, 1 KIT
Catalog # K0492
Plasmid Maxiprep kit, GeneJET™
Catalog # K0492
CAS Number:  
Plasmid Maxiprep kit, GeneJET™
Plasmid Maxiprep kit, GeneJET™

GeneJET™ Plasmid Maxiprep Kit, 25 preps, 1 KIT

CAS Number:  
Price
SFr. 644.00
Log in for availability
1 * 1 KIT
Product Details & Documents

GeneJET™ Plasmid Maxiprep Kit is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. GeneJET™ Plasmid Maxiprep Kit utilises silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 750 µg of high copy plasmid DNA that can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.

  • Efficient: High yields up to 750 µg of high quality plasmid DNA from 250 ml of bacterial culture
  • High concentration of DNA purified: >400 ng/µl
  • Convenient: Spin column format, centrifuge or vacuum protocols
  • Fast: Procedure takes 60 minutes, considerably faster than using gravity flow columns

 : Each kit includes resuspension solution, lysis solution, neutralisation solution, endotoxin binding reagent, Wash Solution I (concentrated), Wash Solution II (concentrated), RNase A solution, elution buffer (10 mM Tris-HCl, pH 8,5), GeneJET™ Maxi purification columns pre-assembled with collection tubes (50 ml), and collection tubes (50 ml).

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    GeneJET™ Plasmid Maxiprep Kit, 25 preps
  • Cat. No.:
    K0492
  • Prep size:
    Maxi
  • Refine target molecule:
    Plasmid
  • Target molecule:
    Plasmid
  • Method / Format:
    Column (silica-based)
  • Description:
    GeneJET™ Plasmid Maxiprep Kit
  • Sample type:
    Bacteria

Specifications