Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Screw top vials

Vials, 15,00 ml, 21×70 mm, Clear, No, 18-400
Catalog # SIAL27162
Supplier:   Merck
Screw top vials
Catalog # SIAL27162
Supplier:  Merck
Screw top vials

Vials, 15,00 ml, 21×70 mm, Clear, No, 18-400

Price
152.00 SFr.
Log in for availability
1 * 100 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifications
  • Cap included:
    No
  • Cap Size:
    18-400
  • Ø×H:
    21×70 mm
  • Capacity:
    15,00 ml
  • Colour:
    Clear
  • Description:
    Vials