Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Screw top vials

Screw top vials

Vials, graduated, 22,00 ml, 23×85 mm, Clear, No, 20-400
Catalog # SIAL27027-U
Supplier:   Merck
Screw top vials
Catalog # SIAL27027-U
Supplier:  Merck
Screw top vials

Vials, graduated, 22,00 ml, 23×85 mm, Clear, No, 20-400

Price
SFr. 205.00
Log in for availability
1 * 100 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifications
  • Cap included:
    No
  • Cap Size:
    20-400
  • Ø×H:
    23×85 mm
  • Capacity:
    22,00 ml
  • Colour:
    Clear
  • Description:
    Vials, graduated
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.