7-Hydroxyindole
Glass bottle for solids, 5 g
Catalog # COBBIN-0037-5G
Marketsource
Supplier: COMBI-BLOCKS
Jump to:
Product Details & Documents
- CAS Number:2380-84-9
- Formula:C₈H₇NO
- MDL Number:MFCD00152102
- Molecular Weight:133.15 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



