Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Solid phase extraction cartridges, HyperSep™ Diol

Cartridge, Diol, 40 - 60 µm, 3 ml, 200 mg
  HYPE60108-573
Solid phase extraction cartridges, HyperSep™ Diol
  HYPE60108-573
Solid phase extraction cartridges, HyperSep™ Diol
Solid phase extraction cartridges, HyperSep™ Diol

Cartridge, Diol, 40 - 60 µm, 3 ml, 200 mg

 
361.00 SFr.
   
1 * 50 ST
 

A short alkyl chains with polar functional groups are ideal for the isolation of polar compounds.

  • Normal phase extraction of polar compounds
  • Purification of polar compounds

HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customisable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.

Packaging: HyperSep-96 are available either in packs of 100 individual wells or in pre-assembled plates.

 
  • Column Format:
    Cartridge
  • Particle Size:
    40 - 60 µm
  • Capacity:
    3 ml
  • Seq:
    03
  • Bed weight:
    200 mg
  • Phase:
    Diol