Cover glasses
High precision cover glasses, 24×50 mm, #1,5, Borosilicate glass
Catalog # 630-2187
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Cover slides suitable for fluorescence microscopy.
- Borosilicate glass D 263™ M of hydrolytic class 1
Recommended for objectives with high numerical aperture and resolution.
Documents
Certificates
Search by Lot NumberSpecifications
- No.:#1,5
- Description:High precision cover glasses
- L×W:24×50 mm
- Material:Borosilicate glass
- Seq:03