Orders LinkContactUsLinkComponent
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
IV bar kit for supporting intravenous solution bottles and bags, incl. 21.2" long stainless steel bar, mounting hardware and four snap hooks; requires Shelf Kit 5235000 or 5235001 (not included)
Catalog # LBCP5242100
Supplier:   LABCONCO
Accessories for glove boxes, Precise™
Catalog # LBCP5242100
Supplier:  LABCONCO
Accessories for glove boxes, Precise™
Accessories for glove boxes, Precise™
IV bar kit for supporting intravenous solution bottles and bags, incl. 21.2" long stainless steel bar, mounting hardware and four snap hooks; requires Shelf Kit 5235000 or 5235001 (not included)
Price
SFr. 189.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    IV bar kit for supporting intravenous solution bottles and bags, incl. 21.2" long stainless steel bar, mounting hardware and four snap hooks; requires Shelf Kit 5235000 or 5235001 (not included)
  • Seq:
    45