Accessories for glove boxes, Precise™
HEPA-filtered pressure relief vent kit, with 99,99% efficient HEPA filter; for ducting to the outside or room; may be used with 5241200 carbon-filtered vent kit
Katalognr. LBCP5238500
Anbieter: LABCONCO
Direkt zu
Product Details & Documents
Technische Daten
- Description:HEPA-filtered pressure relief vent kit, with 99,99% efficient HEPA filter; for ducting to the outside or room; may be used with 5241200 carbon-filtered vent kit
- Seq:11